siRNA Details
Gene Symbol: 2810439L12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013270.1 | - | 1 | 229 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB797663.1 | - | 1 | 107 | Chr. 4 - 99655717 - 99655896 [-] on Build GRCm38 |
