siRNA Details
Gene Symbol: 2210411M09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK008903.1 | - | 2 | 217 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY708799.1 | - | 2 | 217 | Chr. 1 - 180952062 - 180958149 [-] on Build GRCm38 |
