siRNA Details
Gene Symbol: 4930518I17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015828.1 | - | 1 | 625 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715371.1 | - | 1 | 625 | Chr. 15 - 32226385 - 32227040 [+] on Build GRCm38 |
