siRNA Details
Gene Symbol: Gm11482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077143.1 | - | 4 | 461 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB617036.1 | - | 4 | 460 | Chr. 4 - 63634421 - 63639361 [+] on Build GRCm38 |
