siRNA Details
Gene Symbol: Gm26550 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK136710.1 | - | 3 | 338 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721106.1 | - | 3 | 333 | Chr. 7 - 24631640 - 24637911 [-] on Build GRCm38 |
