siRNA Details
Gene Symbol: 9530051G07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040272.1 | 4 | 821 | |||
GenBank mRNA | AK032014.1 | - | 3 | 523 | ||
AK035460.1 | - | 4 | 822 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB622218.1 | - | 3 | 523 | Chr. X - 153038671 - 153069413 [+] on Build GRCm38 |
