siRNA Details
Gene Symbol: 9530051G07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040272.1 | 2 | 149 | |||
GenBank mRNA | AK035460.1 | - | 2 | 150 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB627959.1 | - | 2 | 144 | Chr. X - 153037634 - 153069170 [+] on Build GRCm38 |
