siRNA Details
Gene Symbol: 5830435N17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017971.1 | - | 1 | 568 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY717885.1 | - | 1 | 564 | Chr. 11 - 96610744 - 96611339 [+] on Build GRCm38 |
