siRNA Details
Gene Symbol: Gm30127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131720.1 | - | 2 | 441 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706382.1 | - | 2 | 441 | Chr. 13 - 35959157 - 35965008 [-] on Build GRCm38 |
