siRNA Details
Gene Symbol: Gm26908 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK134715.1 | - | 120 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ049972.1 | - | 1 | 120 | Chr. 14 - 70205324 - 70208835 [+] on Build GRCm38 |
