siRNA Details
Gene Symbol: C430002N11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102293.1 | 4, 5 | 428 | |||
NR_102296.1 | 4, 5 | 515 | ||||
GenBank mRNA | AA030139.1 | - | 4, 5 | 389 | ||
AK049386.1 | - | 4, 5 | 503 | |||
AK089981.1 | - | 502 |
