siRNA Details
Gene Symbol: C030037D09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038058.1 | 3 | 617 | |||
GenBank mRNA | AK021127.1 | - | 3 | 425 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY728261.1 | - | 3 | 418 | Chr. 11 - 88718842 - 88728363 [+] on Build GRCm38 |
