siRNA Details
Gene Symbol: 4930506A18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015712.1 | - | 4 | 360 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715253.1 | - | 4 | 360 | Chr. 8 - 106282286 - 106335648 [+] on Build GRCm38 |
