siRNA Details
Gene Symbol: Gm39139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133159.1 | - | 232 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB616086.1 | - | 3 | 232 | Chr. 8 - 19169540 - 19188015 [-] on Build GRCm38 |
