siRNA Details
Gene Symbol: 4930518J20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005624.1 | - | 1 | 476 | ||
AK015829.1 | - | 1 | 468 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AI595103.1 | - | 1 | 271 | Chr. 1 - 156303697 - 156304200 [+] on Build GRCm38 |
