siRNA Details
Gene Symbol: 1110013I04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK003663.1 | - | 2 | 577 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY703926.1 | - | 2 | 578 | Chr. 8 - 112961448 - 112962560 [+] on Build GRCm38 |
