siRNA Details
Gene Symbol: 1110013H19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK003662.1 | - | 1 | 264 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY703925.1 | - | 1 | 264 | Chr. 15 - 100613173 - 100613843 [-] on Build GRCm38 |
