siRNA Details
Gene Symbol: 4933416M06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045846.2 | 6 | 714 | |||
GenBank mRNA | AK016834.1 | - | 6 | 714 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716508.1 | - | 6 | 710 | Chr. 13 - 101254323 - 101293055 [+] on Build GRCm38 |
