siRNA Details
Gene Symbol: 4833413N01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014692.1 | - | 1 | 335 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714189.1 | - | 1 | 331 | Chr. 1 - 132078980 - 132079804 [+] on Build GRCm38 |
