siRNA Details
Gene Symbol: 2310061D13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009996.1 | - | 1 | 135 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY709562.1 | - | 1 | 132 | Chr. 12 - 79493258 - 79493635 [-] on Build GRCm38 |
