siRNA Details
Gene Symbol: 2610005G05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011318.1 | - | 1 | 652 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV111739.1 | - | 1 | 263 | Chr. 16 - 26992291 - 26992571 [+] on Build GRCm38 |
