siRNA Details
Gene Symbol: 4930583P06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040612.1 | 3 | 564 | |||
GenBank mRNA | AK016345.1 | - | 3 | 536 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA465385.1 | - | 3 | 567 | Chr. 2 - 124217689 - 124222360 [-] on Build GRCm38 |
