siRNA Details
Gene Symbol: Gm38901 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039590.1 | - | 219 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB633641.1 | - | 1 | 215 | Chr. 6 - 127875931 - 127877243 [-] on Build GRCm38 |
