siRNA Details
Gene Symbol: Gm29674 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK043688.1 | - | 1 | 3694 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB265390.2 | - | 1 | 297 | Chr. 10 - 107200816 - 107202178 [-] on Build GRCm38 |
