siRNA Details
Gene Symbol: 4930412L05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015122.1 | - | 5 | 501 | ||
AK141187.1 | - | 8 | 3865 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714668.1 | - | 5 | 501 | Chr. 4 - 53207279 - 53217561 [-] on Build GRCm38 |
