siRNA Details
Gene Symbol: 4933406J10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046004.1 | 4 | 584 | |||
GenBank mRNA | AK016697.1 | - | 4 | 585 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU945775.1 | - | 3 | 321 | Chr. 7 - 82743357 - 82754626 [-] on Build GRCm38 |
