siRNA Details
Gene Symbol: 4930557F08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016160.1 | - | 4 | 339 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715764.1 | - | 4 | 335 | Chr. 15 - 15779996 - 15823426 [+] on Build GRCm38 |
