siRNA Details
Gene Symbol: Gm20750 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040555.1 | 5 | 3029 | |||
GenBank mRNA | AK034478.1 | - | 5 | 3029 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm20750 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040555.1 | 5 | 3029 | |||
GenBank mRNA | AK034478.1 | - | 5 | 3029 |