siRNA Details
Gene Symbol: Gm20750 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040555.1 | 2 | 126 | |||
GenBank mRNA | AK034478.1 | - | 2 | 126 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB626530.1 | - | 2 | 123 | Chr. 3 - 52909633 - 52927663 [-] on Build GRCm38 |
