siRNA Details
Gene Symbol: Gm15520 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077358.1 | - | 6 | 1155 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB027381.2 | - | 3 | 629 | Chr. 9 - 29065802 - 29070198 [+] on Build GRCm38 |
