siRNA Details
Gene Symbol: 9430007M09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020407.1 | - | 1 | 512 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721578.1 | - | 1 | 511 | Chr. 5 - 135264326 - 135264958 [+] on Build GRCm38 |
