siRNA Details
Gene Symbol: 9430006E15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020405.1 | - | 1 | 221 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721574.1 | - | 1 | 219 | Chr. 9 - 20687820 - 20688466 [+] on Build GRCm38 |
