siRNA Details
Gene Symbol: 1700021L22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006219.1 | - | 1 | 458 | ||
BC048567.1 | - | 1 | 433 | |||
BC099561.1 | - | 1 | 268 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706305.1 | - | 1 | 457 | Chr. 6 - 38433108 - 38433783 [-] on Build GRCm38 |
