siRNA Details
Gene Symbol: Gm35618 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133439.1 | - | 23 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ047871.1 | - | 1 | 23 | Chr. 13 - 69681940 - 69697676 [-] on Build GRCm38 |
