siRNA Details
Gene Symbol: Gm830 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK157456.1 | - | 545 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY752306.1 | - | 3 | 542 | Chr. 4 - 95467547 - 95487392 [-] on Build GRCm38 |
