siRNA Details
Gene Symbol: 9530047P18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020597.1 | - | 2 | 975 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB114905.1 | - | 1 | 108 | Chr. 14 - 65466835 - 65467163 [+] on Build GRCm38 |
