siRNA Details
Gene Symbol: Gm31819 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK141885.1 | - | 2 | 102 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB655980.1 | - | 2 | 102 | Chr. 18 - 72702801 - 72725067 [-] on Build GRCm38 |
