siRNA Details
Gene Symbol: 4921511C10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045502.1 | 2, 3 | 1015 | |||
GenBank mRNA | AK014864.1 | - | 2, 3 | 1015 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4921511C10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045502.1 | 2, 3 | 1015 | |||
GenBank mRNA | AK014864.1 | - | 2, 3 | 1015 |