siRNA Details
Gene Symbol: 9130227L01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033706.1 | - | 1 | 40 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB625327.1 | - | 1 | 40 | Chr. 1 - 55960914 - 55971397 [+] on Build GRCm38 |
