siRNA Details
Gene Symbol: Gm16793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040376.1 | 2 | 424 | |||
GenBank mRNA | AK040224.1 | - | 2 | 377 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB635666.1 | - | 2 | 377 | Chr. 8 - 35583547 - 35588972 [-] on Build GRCm38 |
