siRNA Details
Gene Symbol: 2310020H05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045495.1 | 6 | 594 | |||
GenBank mRNA | AK009418.1 | - | 6 | 596 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2310020H05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045495.1 | 6 | 594 | |||
GenBank mRNA | AK009418.1 | - | 6 | 596 |