siRNA Details
Gene Symbol: A930031G03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020913.1 | - | 1 | 672 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CN700646.1 | - | 1 | 422 | Chr. 15 - 85418236 - 85418804 [+] on Build GRCm38 |
