siRNA Details
Gene Symbol: C330011F03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046166.1 | 5 | 1668 | |||
GenBank mRNA | AK049176.1 | - | 5 | 1668 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C330011F03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046166.1 | 5 | 1668 | |||
GenBank mRNA | AK049176.1 | - | 5 | 1668 |