siRNA Details
Gene Symbol: C330011F03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046166.1 | 2 | 99 | |||
GenBank mRNA | AK049176.1 | - | 2 | 99 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB651731.1 | - | 2 | 99 | Chr. 17 - 51434083 - 51448269 [-] on Build GRCm38 |
