siRNA Details
Gene Symbol: Pantr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110495.1 | 4 | 423 | |||
GenBank mRNA | AK011788.1 | - | 1 | 952 | ||
AK011913.1 | - | 1 | 952 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pantr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110495.1 | 4 | 423 | |||
GenBank mRNA | AK011788.1 | - | 1 | 952 | ||
AK011913.1 | - | 1 | 952 |