siRNA Details
Gene Symbol: Gm9806 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015803.1 | - | 1 | 1156 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU937106.1 | - | 1 | 565 | Chr. 19 - 16781141 - 16781908 [+] on Build GRCm38 |
