siRNA Details
Gene Symbol: 1700066O22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015800.1 | - | 3 | 467 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715343.1 | - | 3 | 464 | Chr. 18 - 57530433 - 57533721 [-] on Build GRCm38 |
