siRNA Details
Gene Symbol: Gm19935 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077163.1 | - | 2 | 453 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB617080.1 | - | 2 | 453 | Chr. 8 - 90816448 - 90817354 [-] on Build GRCm38 |
