siRNA Details
Gene Symbol: 4930549C15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016075.1 | - | 1 | 161 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715674.1 | - | 1 | 156 | Chr. 12 - 26346717 - 26371263 [+] on Build GRCm38 |
