siRNA Details
Gene Symbol: Gm12436 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK048554.1 | - | 1, 2 | 172 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB649414.1 | - | 1, 2 | 172 | Chr. 4 - 48898593 - 48960905 [+] on Build GRCm38 |
